• USA Home
  • 387517 - 2-(4-Aminobenzoyl)-2′-chloroacetanilide

EMAIL THIS PAGE TO A FRIEND
387517 Sigma-Aldrich

2-(4-Aminobenzoyl)-2′-chloroacetanilide

95%

Purchase

Properties

assay   95%
mp   160 °C (dec.) (lit.)
SMILES string   Nc1ccc(cc1)C(=O)CC(=O)Nc2ccccc2Cl
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?