• USA Home
  • 387673 - Polybutadiene, epoxy functionalized, hydroxy terminated

EMAIL THIS PAGE TO A FRIEND
387673 Aldrich

Polybutadiene, epoxy functionalized, hydroxy terminated

average Mw ~2,600, average Mn ~1,300

Purchase

Properties

Related Categories Dienes, Hydrophobic Polymers, Materials Science, Polymer Science, Polymers More...
mol wt   average Mn ~1,300
  average Mw ~2,600
composition   cis-1,4, 15%
  trans-1,4, 55%
  vinyl, 30%
density   1.01 g/mL at 25 °C

Description

Packaging

500 mL in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?