• USA Home
  • 388556 - Ammonium tetrachlorocuprate(II) dihydrate

EMAIL THIS PAGE TO A FRIEND
388556 Sigma-Aldrich

Ammonium tetrachlorocuprate(II) dihydrate

98%

Synonym: Ammonium chlorocuprate(II), Ammonium copper chloride

Purchase

Properties

assay   98%
SMILES string   [NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl
InChI   1S/4ClH.Cu.2H3N.2H2O/h4*1H;;2*1H3;2*1H2/q;;;;+2;;;;/p-2
InChI key   LLMDTWKBBWCWNN-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

12801 Ammonium tetrachlorocuprate(II) dihydrate, purum, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?