• USA Home
  • 38938 - Tributylmethylammonium methyl sulfate

EMAIL THIS PAGE TO A FRIEND
38938 Sigma-Aldrich

Tributylmethylammonium methyl sulfate

≥95%

Synonym: Methyltributylammonium methyl sulfate

Purchase

Properties

Related Categories Ammonium Salts, Ammonium ionic liquids, Chemical Synthesis, Ionic Liquids, Phase Transfer Catalysts,
assay   ≥95%
impurities   ≤0.5% water
anion traces   chloride (Cl-): ≤30 mg/kg
SMILES string   COS([O-])(=O)=O.CCCC[N+](C)(CCCC)CCCC
InChI   1S/C13H30N.CH4O4S/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;1-5-6(2,3)4/h5-13H2,1-4H3;1H3,(H,2,3,4)/q+1;/p-1
InChI key   FIMHASWLGDDANN-UHFFFAOYSA-M

Description

Application

Basionic Ionic Liquids

Packaging

100 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
354.2 °F
Flash Point(C) 
179 °C
Protocols & Articles

Articles

ChemFiles Volume 4 Article 9 - Ionic Liquids

Ionic Liquids have experienced a comet-like boost in the last few years. This fascinating class of new materials with its unusual and extraordinary properties has found innumerable new applications i...
ChemFiles Volume 4 Article 9
Keywords: Addition reactions, Applications, Industries, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?