• USA Home
  • 389552 - 2,2′-Methylenebis(1,3-cyclohexanedione)

EMAIL THIS PAGE TO A FRIEND
389552 Sigma-Aldrich

2,2′-Methylenebis(1,3-cyclohexanedione)

98%

Purchase

Properties

assay   98%
SMILES string   O=C1CCCC(=O)C1CC2C(=O)CCCC2=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?