• USA Home
  • 389706 - Ethyl 2-(octadecylthio)benzoate

EMAIL THIS PAGE TO A FRIEND
389706 Aldrich

Ethyl 2-(octadecylthio)benzoate

99%

  • CAS Number 74010-84-7

  • Linear Formula CH3(CH2)17SC6H4CO2C2H5

  • Molecular Weight 434.72

  •  MDL number MFCD00157258

Purchase

Properties

assay   99%
mp   58-60 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?