• USA Home
  • 390259 - Sodium octyl sulfate

EMAIL THIS PAGE TO A FRIEND
390259 Sigma-Aldrich

Sodium octyl sulfate

95%

Synonym: Octyl sulfate sodium salt

  • CAS Number 142-31-4

  • Linear Formula CH3(CH2)7OSO3Na

  • Molecular Weight 232.27

  •  Beilstein/REAXYS Number 3658635

  •  EC Number 205-535-5

  •  MDL number MFCD00007470

Purchase

Properties

InChI Key   WFRKJMRGXGWHBM-UHFFFAOYSA-M
assay   95%
mp   195 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WT1175000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

O4003 Sodium octyl sulfate, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?