• USA Home
  • 390658 - Bis[3-(heptafluoropropylhydroxymethylene)-(+)-camphorato]oxovanadium(IV)

EMAIL THIS PAGE TO A FRIEND
390658 Sigma-Aldrich

Bis[3-(heptafluoropropylhydroxymethylene)-(+)-camphorato]oxovanadium(IV)

98%

  • CAS Number 130552-91-9

  • Empirical Formula (Hill Notation) C28H28F14O5V

  • Molecular Weight 761.44

  •  MDL number MFCD00191812

Purchase

Properties

assay   98%
optical activity   [α]20/D +224°, c = 0.06 in methylene chloride
mp   140 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?