• USA Home
  • 390771 - 2,4,4-Trimethylcyclopentanone

EMAIL THIS PAGE TO A FRIEND
390771 Sigma-Aldrich

2,4,4-Trimethylcyclopentanone

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.432 (lit.)
bp   160 °C (lit.)
density   0.877 g/mL at 25 °C (lit.)
SMILES string   CC1CC(C)(C)CC1=O
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN1224 - class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?