• USA Home
  • 392316 - 2-(Benzylmethylamino)-4′-hydroxy-3′-methoxyacetophenone hydrochloride

EMAIL THIS PAGE TO A FRIEND
392316 Sigma-Aldrich

2-(Benzylmethylamino)-4′-hydroxy-3′-methoxyacetophenone hydrochloride

  • CAS Number 206752-44-5

  • Linear Formula HOC6H3(OCH3)COCH2N(CH3)CH2C6H5·HCl

  • Molecular Weight 321.80

  •  MDL number MFCD00191777

  •  PubChem Substance ID 24864443

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
mp   190-200 °C (lit.)
SMILES string   Cl.COc1cc(ccc1O)C(=O)CN(C)Cc2ccccc2
InChI   1S/C17H19NO3.ClH/c1-18(11-13-6-4-3-5-7-13)12-16(20)14-8-9-15(19)17(10-14)21-2;/h3-10,19H,11-12H2,1-2H3;1H
InChI key   GHCRKQYZNMBJLO-UHFFFAOYSA-N

Description

General description

2-(Benzylmethylamino)-4′-hydroxy-3′-methoxyacetophenone hydrochloride is an acetophenone derivative.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?