• USA Home
  • 392812 - trans-3-Benzyloxy-4-methoxy-β-nitrostyrene

EMAIL THIS PAGE TO A FRIEND
392812 Sigma-Aldrich

trans-3-Benzyloxy-4-methoxy-β-nitrostyrene

99%

Purchase

Properties

assay   99%
mp   128-131 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?