• USA Home
  • 393886 - Copper(II) 3,10,17,24-tetra-tert-butyl-1,8,15,22-tetrakis(dimethylamino)-29H,31H-phthalocyanine

EMAIL THIS PAGE TO A FRIEND
393886 Sigma-Aldrich

Copper(II) 3,10,17,24-tetra-tert-butyl-1,8,15,22-tetrakis(dimethylamino)-29H,31H-phthalocyanine

Dye content 80 %

  • CAS Number 61113-98-2

  • Empirical Formula (Hill Notation) C56H68CuN12

  • Molecular Weight 972.77

  •  MDL number MFCD00191952

  •  PubChem Substance ID 24864547

Purchase

Properties

composition   Dye content, 80%
absorption   λmax 768 nm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?