• USA Home
  • 394033 - 3-Amino-1-(2,4,6-trichlorophenyl)-2-pyrazolin-5-one

EMAIL THIS PAGE TO A FRIEND
394033 Sigma-Aldrich

3-Amino-1-(2,4,6-trichlorophenyl)-2-pyrazolin-5-one

97%

Purchase

Properties

assay   97%
mp   218 °C (dec.) (lit.)
SMILES string   NC1=NN(C(=O)C1)c2c(Cl)cc(Cl)cc2Cl
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?