• USA Home
  • 394068 - 5-[4,5-Dihydro-5-oxo-3-[(1-oxooctadecyl)amino]-1H-pyrazol-1-yl]-2-phenoxybenzenesulfonic acid

EMAIL THIS PAGE TO A FRIEND
394068 Sigma-Aldrich

5-[4,5-Dihydro-5-oxo-3-[(1-oxooctadecyl)amino]-1H-pyrazol-1-yl]-2-phenoxybenzenesulfonic acid

97%

Purchase

Properties

assay   97%
mp   237-240 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCCCC(=O)NC1=NN(C(=O)C1)c2ccc(Oc3ccccc3)c(c2)S(O)(=O)=O
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2585PSN2 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?