EMAIL THIS PAGE TO A FRIEND
394793 Sigma-Aldrich

Barium hexaboride

−100 mesh

Purchase

Properties

particle size   −100 mesh
density   4.36 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1564 6.1 / PGII
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?