• USA Home
  • 395412 - Hafnium(IV) chloride tetrahydrofuran complex (1:2)

EMAIL THIS PAGE TO A FRIEND
395412 Sigma-Aldrich

Hafnium(IV) chloride tetrahydrofuran complex (1:2)

98%

Synonym: Tetrachlorobis(tetrahydrofuran)hafnium

Purchase

Properties

Related Categories Essential Chemicals, Hafnium Salts, Materials Science, Metal and Ceramic Science, Research Essentials,
InChI Key   KSIFQVHSGOEHQQ-UHFFFAOYSA-J
assay   98%
form   powder
mp   187 °C (lit.)
storage temp.   2-8°C

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2925 8(4.1) / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?