• USA Home
  • 395447 - Methyl(3,3,3-trifluoropropyl)cyclopolysiloxanes, mixture of tri- and tetra-

EMAIL THIS PAGE TO A FRIEND
395447 Sigma-Aldrich

Methyl(3,3,3-trifluoropropyl)cyclopolysiloxanes, mixture of tri- and tetra-

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.371 (lit.)
bp   95-134 °C/3 mmHg (lit.)
mp   31-33 °C (lit.)
density   1.264 g/mL at 25 °C (lit.)
SMILES string   C[Si](=O)CCC(F)(F)F
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?