• USA Home
  • 396214 - 3-Chlorododecanedioic acid

EMAIL THIS PAGE TO A FRIEND
396214 Sigma-Aldrich

3-Chlorododecanedioic acid

98%

  • CAS Number 37443-68-8

  • Linear Formula HO2C(CH2)8CH(Cl)CH2CO2H

  • Molecular Weight 264.75

  •  MDL number MFCD00191916

Purchase

Properties

assay   98%
mp   107-109 °C (lit.)
SMILES string   OC(=O)CCCCCCCCC(Cl)CC(O)=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?