• USA Home
  • 39927 - cis-2-Amino-2-methylcyclopentanecarboxylic acid hydrochloride

EMAIL THIS PAGE TO A FRIEND
39927 Sigma-Aldrich

cis-2-Amino-2-methylcyclopentanecarboxylic acid hydrochloride

≥98.0% (CHN)

  • CAS Number 156292-34-1

  • Empirical Formula (Hill Notation) C7H13NO2 · HCl

  • Molecular Weight 179.64

  •  MDL number MFCD05863639

  •  PubChem Substance ID 57649912

  •  NACRES NA.22

Purchase

Properties

Related Categories Alicyclic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   VUBCCMLFYBOWSD-KQBMADMWSA-N
assay   ≥98.0% (CHN)

Description

Packaging

500 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Unnatural Amino Acids - ChemFiles 2008, 8.7, 3.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research. D...
Matthias Junkers
ChemFiles 2008, 8.7, 3.
Keywords: Absorption, Addition reactions, Alkylations, Asymmetric synthesis, Building blocks, Degradations, Drug discovery, Metabolism, Molecular probes, Peptidomimetics, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?