• USA Home
  • 399345 - 2-(Trifluoroacetoxy)pyridine

EMAIL THIS PAGE TO A FRIEND
399345 Sigma-Aldrich

2-(Trifluoroacetoxy)pyridine

98%

Synonym: 2-Pyridyl trifluoroacetate

  • CAS Number 96254-05-6

  • Empirical Formula (Hill Notation) C7H4F3NO2

  • Molecular Weight 191.11

  •  Beilstein/REAXYS Number 3607316

  •  MDL number MFCD00192522

Purchase

Properties

InChI Key   BJCXMOBNIFBHBJ-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.421 (lit.)
bp   60 °C/0.3 mmHg(lit.)
density   1.356 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

91671 2-(Trifluoroacetoxy)pyridine, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?