• USA Home
  • 399639 - 5,6-Dimethyl-2,3-pyrazinedicarbonitrile

EMAIL THIS PAGE TO A FRIEND
399639 Sigma-Aldrich

5,6-Dimethyl-2,3-pyrazinedicarbonitrile

99%

  • CAS Number 40227-17-6

  • Empirical Formula (Hill Notation) C8H6N4

  • Molecular Weight 158.16

  •  MDL number MFCD00192087

  •  PubChem Substance ID 24864831

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazines
InChI Key   VPRRXMCIOHZBCX-UHFFFAOYSA-N
assay   99%
mp   168-170 °C (dec.) (lit.)

Description

General description

5,6-Dimethyl-2,3-pyrazinedicarbonitrile, a pyrazine (paradiazine) derivative, is a heterocyclic building block. It is a heterocyclic six-membered aromatic compound bearing nitrogen atoms at para positions.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?