• USA Home
  • 400041 - Methyl 9-fluorenylideneacetate

EMAIL THIS PAGE TO A FRIEND
400041 Sigma-Aldrich

Methyl 9-fluorenylideneacetate

95%

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   JKKWDADOWDCWKC-UHFFFAOYSA-N
assay   95%
mp   108-112 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?