• USA Home
  • 400254 - Lithium ethoxide solution

EMAIL THIS PAGE TO A FRIEND
400254 Sigma-Aldrich

Lithium ethoxide solution

1.0 M in THF

  • CAS Number 2388-07-0

  • Linear Formula CH3CH2OLi

  • Molecular Weight 52.00

  •  Beilstein/REAXYS Number 3909950

  •  MDL number MFCD00050493

  •  PubChem Substance ID 24864907

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Lithium, Materials Science,
InChI Key   AZVCGYPLLBEUNV-UHFFFAOYSA-N
form   liquid
concentration   1.0 M in THF
bp   67 °C
density   0.895 g/mL at 25 °C

Description

Packaging

100 mL in Sure/Seal™

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides., Reacts violently with water.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
-20.2 °F
Flash Point(C) 
-29 °C
Protocols & Articles

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Department of Mechanical Engineering and Materials Science, One Brookings Drive, Washington University in St. Louis, St. Louis, MO 63130 *Email: parag.banerjee@wustl.edu
Keywords: Atomic layer deposition, Catalysis, Chemical reactions, Degradations, Dehydrogenation, Deposition, Hydrogenations, Ion Exchange, Ligands, Magnetic resonance spectroscopy, Materials Science, Nanomaterials, Oxidations, Photovoltaics, Precipitation, Recombination, Reductions, Renewable energy, Separation, Solar cells, Thin film deposition

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?