• USA Home
  • 401099 - Bismuth(III) telluride

EMAIL THIS PAGE TO A FRIEND
401099 Aldrich

Bismuth(III) telluride

Synonym: Bismuth sesquitelluride, Bismuth telluride, Dibismuth tritelluride

Purchase

Properties

InChI Key   GUYIRKJSQUOSJV-UHFFFAOYSA-N
mp   585 °C (lit.)
density   7.642 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN3284 - class 6.1 - PG 3 - MP - Tellurium compound, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
EB3110000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?