EMAIL THIS PAGE TO A FRIEND
40215 Sigma-Aldrich

Hexamethyldisilazane

semiconductor grade PURANAL (Honeywell 17713)

Synonym: HMDS

  • CAS Number 999-97-3

  • Linear Formula (CH3)3SiNHSi(CH3)3

  • Molecular Weight 161.39

  •  Beilstein/REAXYS Number 635752

  •  EC Number 213-668-5

  •  MDL number MFCD00008259

  •  PubChem Substance ID 57649936

Purchase

Properties

Related Categories Chemical Synthesis, Electronic Chemicals, Materials Science, Materials for Surface Modification, Micro/NanoElectronics,
grade   semiconductor grade PURANAL (Honeywell 17713)
InChI Key   FFUAGWLWBBFQJT-UHFFFAOYSA-N
assay   99.70%
CofA   specification on request
refractive index   n20/D 1.407 (lit.)
bp   125 °C (lit.)

Description

General description

Hexamethyldisilazane (HDMS) is a bulk organo silicon compound. It is hydrophobic in nature.It finds major application as a silanizing agent, silylation with HMDS is carried out in the presence of an acid catalyst.The hydrophobic nature of HDMS coatings on various nanoparticles renders them resistant to water contamination and flocculations, during synthesis.

Application

HDMS was used to silanize the surface of silicon wafer,cellulose. HDMS may be used to dehydrate cells of biomaterials for scanning electron microscopy (SEM). HDMS mediated ring opening polymerization of Α-helical polypeptides. Zinc blende nanocrystals were prepared in the presence of HDMS.

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1992 6.1(3) / PGII
WGK Germany 
2
RTECS 
JM9230000
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?