EMAIL THIS PAGE TO A FRIEND
40219 Sigma-Aldrich

2-Propanol

semiconductor grade PURANAL (Honeywell 17829)

Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

  • CAS Number 67-63-0

  • Linear Formula (CH3)2CHOH

  • Molecular Weight 60.10

  •  Beilstein Registry Number 635639

  •  EC Number 200-661-7

  •  MDL number MFCD00011674

  •  PubChem Substance ID 57649938

Purchase

Properties

Related Categories Electronic Chemicals, Materials Science, Micro/NanoElectronics, Semiconductor Grade Chemicals, Semiconductor Solvents,
grade   semiconductor grade PURANAL (Honeywell 17829)
vapor density   2.1 (vs air)
vapor pressure   33 mmHg ( 20 °C)
  44 mmHg ( 25 °C)
InChI Key   KFZMGEQAYNKOFK-UHFFFAOYSA-N
form   liquid
CofA   specification on request
autoignition temp.   750 °F
expl. lim.   2 %
color   colorless
refractive index   n20/D 1.377(lit.)
  n/D
bp   82 °C(lit.)
mp   −89.5 °C (lit.)
solubility   water: soluble (completely)
density   0.785 g/mL at 25 °C(lit.)

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1219 - class 3 - PG 2 - Isopropanol
WGK Germany 
1
RTECS 
NT8050000
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

Use the interactive controls above to simplify calculations and improve the efficiency of your distillation or evaporation requirements.
Keywords: Evaporation, Vaporization

Properties of Solvents Table

The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?