• USA Home
  • 40292 - Ammonium fluoride solution

EMAIL THIS PAGE TO A FRIEND
40292 Sigma-Aldrich

Ammonium fluoride solution

semiconductor grade VLSI PURANAL (Honeywell 17600), 40%

Purchase

Properties

Related Categories Chemical Synthesis, Deprotecting Reagents, Electronic Chemicals, Fluoride Sources, Materials Science,
grade   semiconductor grade VLSI PURANAL (Honeywell 17600)
assay   40%
CofA   specification on request
SMILES string   N.F
InChI   1S/FH.H3N/h1H;1H3
InChI key   LDDQLRUQCUTJBB-UHFFFAOYSA-N

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3287 6.1 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?