EMAIL THIS PAGE TO A FRIEND
40343 Aldrich

2-Propanol

semiconductor grade SLSI PURANAL (Honeywell 17030)

Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

  • CAS Number 67-63-0

  • Linear Formula (CH3)2CHOH

  • Molecular Weight 60.10

  •  Beilstein Registry Number 635639

  •  EC Number 200-661-7

  •  MDL number MFCD00011674

  •  PubChem Substance ID 57649987

Purchase

Properties

Related Categories Electronic Chemicals, Materials Science, Micro/NanoElectronics, Semiconductor Grade Chemicals, Semiconductor Solvents,
grade   semiconductor grade SLSI PURANAL (Honeywell 17030)
vapor density   2.1 (vs air)
vapor pressure   33 mmHg ( 20 °C)
  44 mmHg ( 25 °C)
InChI Key   KFZMGEQAYNKOFK-UHFFFAOYSA-N
CofA   specification on request
autoignition temp.   750 °F
expl. lim.   2 %
refractive index   n20/D 1.377(lit.)
bp   82 °C(lit.)
mp   −89.5 °C(lit.)
density   0.785 g/mL at 25 °C(lit.)

Description

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1219 3 / PGII
WGK Germany 
1
RTECS 
NT8050000
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles

Related Content

Pressure-Temperature Nomograph Interactive Tool

The Pressure-Temperature Nomograph tool is an application of the Clausius-Clapeyron Equation, which assumes the heat of vaporization is a constant over a pressure range. Antoine´s Equation gets aroun...
Keywords: Evaporation, Vaporization

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?