• USA Home
  • 403628 - Germanium(IV) chloride solution

EMAIL THIS PAGE TO A FRIEND
403628 Sigma-Aldrich

Germanium(IV) chloride solution

1.0 M in methylene chloride

  • CAS Number 10038-98-9

  • Empirical Formula (Hill Notation) Cl4Ge

  • Molecular Weight 214.45

  •  MDL number MFCD00011029

Purchase

Properties

concentration   1.0 M in methylene chloride
SMILES string   Cl[Ge](Cl)(Cl)Cl

Description

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3390 8(6.1) / PGI
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?