EMAIL THIS PAGE TO A FRIEND
404209 Sigma-Aldrich

Tetrabutylgermanium

94%

Purchase

Properties

assay   94%
refractive index   n20/D 1.455(lit.)
bp   >100 °C(lit.)
density   0.93 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?