• USA Home
  • 404373 - Chromium(0) hexacarbonyl

EMAIL THIS PAGE TO A FRIEND
404373 Aldrich

Chromium(0) hexacarbonyl

technical grade, 96%

Purchase

Properties

grade   technical grade
vapor density   7.6 (vs air)
vapor pressure   1 mmHg ( 36 °C)
InChI Key   KOTQLLUQLXWWDK-UHFFFAOYSA-N
assay   96%
mp   >150 °C (dec.)(lit.)
density   1.77 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 3466 6.1/PG 3
WGK Germany 
3
RTECS 
GB5075000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

241458 Chromium(0) hexacarbonyl, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?