• USA Home
  • 404616 - Hexafluoroacetone azine

EMAIL THIS PAGE TO A FRIEND
404616 Sigma-Aldrich

Hexafluoroacetone azine

99%

  • CAS Number 1619-84-7

  • Linear Formula (CF3)2C=NN=C(CF3)2

  • Molecular Weight 328.06

  •  Beilstein/REAXYS Number 1916266

  •  MDL number MFCD00067583

  •  PubChem Substance ID 24865260

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks, Other Nitrogen Compounds More...
assay   99%
refractive index   n20/D 1.3 (lit.)
bp   67-68 °C (lit.)
density   1.439 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)\C(=N/N=C(/C(F)(F)F)C(F)(F)F)C(F)(F)F
InChI   1S/C6F12N2/c7-3(8,9)1(4(10,11)12)19-20-2(5(13,14)15)6(16,17)18
InChI key   SZLJMGDRBNZIIE-UHFFFAOYSA-N

Description

Packaging

1 g in ampule

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?