EMAIL THIS PAGE TO A FRIEND
405167 Sigma-Aldrich

IR-1051

Dye content 95 %

  • CAS Number 100012-45-1

  • Empirical Formula (Hill Notation) C41H33BCl2F4N2

  • Molecular Weight 711.42

  •  MDL number MFCD00191903

  •  PubChem Substance ID 24865295

Purchase

Properties

Related Categories Infrared (IR) Dyes, Laser Dyes, Materials Science, Organic and Printed Electronics, Photonic and Optical Materials More...
composition   Dye content, 95%
mp   230 °C (dec.) (lit.)
λmax   1051 nm
SMILES string   F[B-](F)(F)F.CCN1\C(=C\C=C2/CCC(\C=C\C3=[N+](CC)c4ccc(Cl)c5cccc3c45)=C2c6ccccc6)c7cccc8c(Cl)ccc1c78
InChI   1S/C41H33Cl2N2.BF4/c1-3-44-35(31-14-8-12-29-33(42)20-24-37(44)40(29)31)22-18-27-16-17-28(39(27)26-10-6-5-7-11-26)19-23-36-32-15-9-13-30-34(43)21-25-38(41(30)32)45(36)4-2;2-1(3,4)5/h5-15,18-25H,3-4,16-17H2,1-2H3;/q+1;-1
InChI key   ARGQPGULWAESKB-UHFFFAOYSA-N

Description

Other Notes

Q-switching IR laser dye. λmaX = 1050nm (C2H5OH)

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?