• USA Home
  • 406260 - Boc-β-iodo-D-Ala-OBzl

EMAIL THIS PAGE TO A FRIEND
406260 Aldrich

Boc-β-iodo-D-Ala-OBzl

99%

Synonym: N-(tert-Butoxycarbonyl)-3-iodo-D-alanine benzyl ester, Boc-3-iodo-D-alanine benzyl ester

  • CAS Number 125942-79-2

  • Linear Formula ICH2CH[NHCO2C(CH3)3]CO2CH2C6H5

  • Molecular Weight 405.23

  •  Beilstein/REAXYS Number 5854502

  •  MDL number MFCD00191867

Purchase

Properties

InChI Key   DDXFSYLOWHQCEK-GFCCVEGCSA-N
assay   99%
optical activity   [α]20/D +18°, c = 1 in ethanol
mp   80-84 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

15122 Boc-β-iodo-D-Ala-OBzl, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?