• USA Home
  • 40678 - Benzoylamidrazone hydrochloride

EMAIL THIS PAGE TO A FRIEND
40678 Sigma-Aldrich

Benzoylamidrazone hydrochloride

technical, ≥90% (HPLC)

Synonym: N-(2-Ethoxybenzoyl)-N′-butyrimidohydrazide hydrochloride

  • CAS Number 437717-21-0

  • Empirical Formula (Hill Notation) C13H19N3O2 · HCl

  • Molecular Weight 285.77

  •  MDL number MFCD03791077

  •  PubChem Substance ID 24865406

Purchase

Properties

grade   technical
assay   ≥90% (HPLC)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?