EMAIL THIS PAGE TO A FRIEND
408638 Sigma-Aldrich

Fmoc-Ile-OH

98%

Synonym: N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine, Fmoc-L-isoleucine

  • CAS Number 71989-23-6

  • Empirical Formula (Hill Notation) C21H23NO4

  • Molecular Weight 353.41

  •  Beilstein/REAXYS Number 4716717

  •  EC Number 276-255-9

  •  MDL number MFCD00037125

Purchase

Properties

InChI Key   QXVFEIPAZSXRGM-DJJJIMSYSA-N
assay   98%
optical activity   [α]20/D −12°, c = 1 in DMF
mp   145-147 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?