EMAIL THIS PAGE TO A FRIEND
408654 Sigma-Aldrich

Fmoc-Asn-OH

98%

Synonym: -(9-Fluorenylmethoxycarbonyl)-L-asparagine, Fmoc-L-asparagine

  • CAS Number 71989-16-7

  • Empirical Formula (Hill Notation) C19H18N2O5

  • Molecular Weight 354.36

  •  Beilstein/REAXYS Number 4335103

  •  EC Number 276-252-2

  •  MDL number MFCD00037132

  •  PubChem Substance ID 24865584

Purchase

Properties

assay   98%
optical activity   [α]20/D −13°, c = 1 in DMF
mp   190 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?