EMAIL THIS PAGE TO A FRIEND
408662 Sigma-Aldrich

Fmoc-Met-OH

97%

Synonym: Fmoc-L-methionine

  • CAS Number 71989-28-1

  • Empirical Formula (Hill Notation) C20H21NO4S

  • Molecular Weight 371.45

  •  Beilstein/REAXYS Number 4300266

  •  EC Number 276-258-5

  •  MDL number MFCD00037134

Purchase

Properties

InChI Key   BUBGAUHBELNDEW-SFHVURJKSA-N
assay   97%
optical activity   [α]20/D −25°, c = 1 in DMF
mp   121-123 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

47634 Fmoc-Met-OH, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?