EMAIL THIS PAGE TO A FRIEND
408689 Sigma-Aldrich

Fmoc-Trp-OH

97%

Synonym: Fmoc-L-tryptophan

  • CAS Number 35737-15-6

  • Empirical Formula (Hill Notation) C26H22N2O4

  • Molecular Weight 426.46

  •  Beilstein/REAXYS Number 4216624

  •  EC Number 252-706-5

  •  MDL number MFCD00037126

  •  PubChem Substance ID 24865587

Purchase

Properties

assay   97%
optical activity   [α]20/D −28°, c = 1 in DMF
impurities   <3% ethyl acetate
mp   182-185 °C (lit.)
SMILES string   OC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCC3c4ccccc4-c5ccccc35
InChI   1S/C26H22N2O4/c29-25(30)24(13-16-14-27-23-12-6-5-7-17(16)23)28-26(31)32-15-22-20-10-3-1-8-18(20)19-9-2-4-11-21(19)22/h1-12,14,22,24,27H,13,15H2,(H,28,31)(H,29,30)/t24-/m0/s1
InChI key   MGHMWKZOLAAOTD-DEOSSOPVSA-N

Description

Application

Cyclic peptides used as tachykinin antagonists have been prepared from Fmoc-protected D- and L- tryptophans.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

47637 Fmoc-Trp-OH, ≥97.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?