• USA Home
  • 410268 - 1-[N-[Poly(3-allyloxy-2-hydroxypropyl)]-2-aminoethyl]-2-imidazolidinone solution

EMAIL THIS PAGE TO A FRIEND
410268 Sigma-Aldrich

1-[N-[Poly(3-allyloxy-2-hydroxypropyl)]-2-aminoethyl]-2-imidazolidinone solution

90 wt. % in H2O

Purchase

Properties

Related Categories Allyl Monomers, Materials Science, Monomers, Polymer Science
concentration   90 wt. % in H2O
refractive index   n20/D 1.494
density   1.15 g/mL at 25 °C

Description

Application

Adhesion promoter

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?