• USA Home
  • 410500 - N-Lithioethylenediamine

EMAIL THIS PAGE TO A FRIEND
410500 Aldrich

N-Lithioethylenediamine

Synonym: Ethylenediamine lithium salt

Purchase
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2813 4.3 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?