• USA Home
  • 411574 - Ethylene glycol bis(propylene glycol-block-ethylene glycol) ether

EMAIL THIS PAGE TO A FRIEND
411574 Aldrich

Ethylene glycol bis(propylene glycol-block-ethylene glycol) ether

average Mn ~2,000

  • CAS Number 53637-25-5

  • Linear Formula HO(CH2CH2O)x(C3H6O)yCH2CH2O(C3H6O)y(CH2CH2O)xH

  •  PubChem Substance ID 24865755

Purchase

Properties

mol wt   average Mn ~2,000
refractive index   n20/D 1.453(lit.)
mp   −32 °C(lit.)
density   1.017 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?