• USA Home
  • 411582 - 1-Chloro-1-methylsilacyclobutane

EMAIL THIS PAGE TO A FRIEND
411582 Aldrich

1-Chloro-1-methylsilacyclobutane

98%

  • CAS Number 2351-34-0

  • Empirical Formula (Hill Notation) C4H9ClSi

  • Molecular Weight 120.65

  •  MDL number MFCD00191926

  •  PubChem Substance ID 24865756

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
InChI Key   JWKKXHBGWAQZOK-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.45(lit.)
bp   103 °C(lit.)
density   0.985 g/mL at 25 °C(lit.)

Description

Application

Intermediate in the formation of enoxysilacyclobutanes.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2985 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
37.4 °F
Flash Point(C) 
3 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?