• USA Home
  • 412139 - Neopentyl glycol ethoxylate (1 EO/OH) diacrylate

EMAIL THIS PAGE TO A FRIEND
412139 Aldrich

Neopentyl glycol ethoxylate (1 EO/OH) diacrylate

average Mn ~300

  • CAS Number 62180-73-8

  • Linear Formula [H2C=CHCO2(CH2CH2O)nCH2]2C(CH3)2

  • Molecular Weight 300.35

  •  MDL number MFCD00192344

Purchase

Properties

vapor pressure   0.02 mmHg ( 25 °C)
mol wt   average Mn ~300
contains   300 ppm monomethyl ether hydroquinone as inhibitor
refractive index   n20/D 1.456 (lit.)
density   1.041 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?