• USA Home
  • 414158 - N-[3-(Trimethoxysilyl)propyl]-N′-(4-vinylbenzyl)ethylenediamine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
414158 Sigma-Aldrich

N-[3-(Trimethoxysilyl)propyl]-N′-(4-vinylbenzyl)ethylenediamine hydrochloride solution

40 wt. % in methanol

  • CAS Number 33401-49-9

  • Linear Formula H2C=CHC6H4CH2NHCH2CH2NH(CH2)3Si(OCH3)3·HCl

  • Molecular Weight 374.98

  •  MDL number MFCD00054284

  •  PubChem Substance ID 24865897

  •  NACRES NA.23

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Monomers, Polymer Science, Self Assembly and Lithography,
Quality Level   100
concentration   40 wt. % in methanol
refractive index   n20/D 1.402
density   0.905 g/mL at 25 °C
SMILES string   Cl.CO[Si](CCCNCCNCc1ccc(C=C)cc1)(OC)OC
InChI   1S/C17H30N2O3Si.ClH/c1-5-16-7-9-17(10-8-16)15-19-13-12-18-11-6-14-23(20-2,21-3)22-4;/h5,7-10,18-19H,1,6,11-15H2,2-4H3;1H
InChI key   JWIKADZFCMEWBV-UHFFFAOYSA-N

Description

Packaging

250 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
WGK 3
Flash Point(F) 
55.4 °F - closed cup
Flash Point(C) 
13 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?