• USA Home
  • 414298 - Bromoacetic anhydride

EMAIL THIS PAGE TO A FRIEND
414298 Sigma-Aldrich

Bromoacetic anhydride

95%

Purchase

Properties

assay   95%
mp   31-36 °C (lit.)
SMILES string   BrCC(=O)OC(=O)CBr
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?