• USA Home
  • 414573 - 2-(p-Toluidino)-6-naphthalenesulfonic acid potassium salt

EMAIL THIS PAGE TO A FRIEND
414573 Sigma-Aldrich

2-(p-Toluidino)-6-naphthalenesulfonic acid potassium salt

technical grade, 85%

Synonym: TNS

Purchase

Properties

grade   technical grade
assay   85%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?