• USA Home
  • 415162 - Barium strontium tungsten oxide

EMAIL THIS PAGE TO A FRIEND
415162 Sigma-Aldrich

Barium strontium tungsten oxide

powder, 10 μm, 99.8% trace metals basis

Synonym: Tungstic acid, barium strontium salt

Purchase

Properties

Related Categories Barium, Materials Science, Metal and Ceramic Science, Oxides
assay   99.8% trace metals basis
form   powder
particle size   10 μm
SMILES string   O=[Sr].O=[Ba].O=[Ba].O=[W](=O)=O
InChI   1S/2Ba.6O.Sr.W
InChI key   ZRFCSKHAMZALKG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?