• USA Home
  • 415391 - 1-(N,N-Dimethylcarbamoyl)-4-(2-sulfoethyl)pyridinium hydroxide inner salt

EMAIL THIS PAGE TO A FRIEND
415391 Aldrich

1-(N,N-Dimethylcarbamoyl)-4-(2-sulfoethyl)pyridinium hydroxide inner salt

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   SJHHXGGXZKPIDE-UHFFFAOYSA-N
mp   185 °C (dec.)(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?