• USA Home
  • 415677 - 2-Nitro-5-(propylthio)aniline

EMAIL THIS PAGE TO A FRIEND
415677 Sigma-Aldrich

2-Nitro-5-(propylthio)aniline

99%

Purchase

Properties

assay   99%
mp   71-74 °C (lit.)
SMILES string   CCCSc1ccc(c(N)c1)[N+]([O-])=O
InChI   1S/C9H12N2O2S/c1-2-5-14-7-3-4-9(11(12)13)8(10)6-7/h3-4,6H,2,5,10H2,1H3
InChI key   NLCPQMUVBQILMA-UHFFFAOYSA-N

Description

General description

2-Nitro-5-(propylthio)aniline is an sulphur containing organic building block.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?